missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DYRK4 Recombinant Human Protein

DYRK4 is a dual-specificity protein kinase.

Brand:  Invitrogen™ PV4189


Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10006512

  • 13340.00 € / 1mg
Estimated Shipment: 20-04-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Dual-specificity kinases, such as DYRK4, play key roles in cell proliferation, survival, and development.

Specifications

Specifications

NP_003836.1
Frozen
88.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
DYRK Kinase Family
Yes
Baculovirus
Protein Biology, Enzyme & Protein Activity Assays, Kinase Activity, Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
DYRK4
Human
GST Tag
Kinase Assay
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers