missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium foil, 0.025mm (0.001in) thick, 99.9% (REO)

Product Code. 11301779
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
Product Code. Quantity unitSize
11301779 25 x 25 mm 25mm
11450270 50 x 50 mm Each
2 options
This item is not returnable. View return policy

Product Code. 11301779

Brand: Thermo Scientific Alfa Aesar 012383.FF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 0.13g/25 x 25mm | CAS: 7429-91-6 | Dy | 162.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-91-6
Molecular Formula Dy
Molecular Weight (g/mol) 162.50
MDL Number MFCD00010982
InChI Key KBQHZAAAGSGFKK-UHFFFAOYSA-N
Synonym ingot reo, disprosio, atom, ion, ingot, pieces, powder, foil, 3n, chips, 3n, ingot, 3n
PubChem CID 23912
ChEBI CHEBI:33377
IUPAC Name dysprosium
SMILES [Dy]

Specifications

Color Gray to Silver
Physical Form Foil
Quantity 25 x 25 mm
Assay Percent Range 99.90%
Odor Odorless
Packaging Under Argon
Appearance Gray to silver foil
Density 8.55g/cm3
Chemical Name or Material Dysprosium, Rare Earth Oxides Content

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.