missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) nitrate pentahydrate, 99.9% (REO)

Product Code. 11337657
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11337657 100 g 100g
11327657 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11337657

Brand: Thermo Scientific Alfa Aesar 012922.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10031-49-9 | DyH10N3O14 | 438.59 g/mol

Dysprosium(III) nitrate pentahydrate is used to prepare carboxylate-based dinuclear dysprosium compound by reacting with n-butyric acid and triethylamine using methanol as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-49-9
Molecular Formula DyH10N3O14
Molecular Weight (g/mol) 438.59
MDL Number MFCD00149683
InChI Key NOTQUFQJAWMLCE-UHFFFAOYSA-N
IUPAC Name dysprosium(3+) pentahydrate trinitrate
SMILES O.O.O.O.O.[Dy+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 88.6°C
Color Yellow
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
Formula Weight 438.59 (348.51 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Dysprosium(III) nitrate pentahydrate, Rare Earth Oxides Content

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.