missing translation for 'onlineSavingsMsg'
Learn More

Dysprosium(III) trifluoromethanesulfonate, 98%

Product Code. 11379106
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379106 5 g 5g
11389106 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379106 Supplier Thermo Scientific Alfa Aesar Supplier No. L20252.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 139177-62-1 | C3DyF9O9S3 | 609.69 g/mol

Dysprosium(III) trifluoromethanesulfonate is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes. This is an effective catalyst for electrophilic substitution reactions of indoles with imines. Also used as catalyst for Aza-Piancatelli rearrangement, Friedel-Crafts alkylation, ring-opening polymerization reactions, microwave-assisted Kabachnik-Fields condensation, cycloaddition reactions (Lewis-acid catalyst), fries rearrangement and enantioselective glyoxalate-ene reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dysprosium(III) trifluoromethanesulfonate is a water-tolerant Lewis acid used in the Aldol reaction of silyl enol ethers with aldehydes. This is an effective catalyst for electrophilic substitution reactions of indoles with imines. Also used as catalyst for Aza-Piancatelli rearrangement, Friedel-Crafts alkylation, ring-opening polymerization reactions, microwave-assisted Kabachnik-Fields condensation, cycloaddition reactions (Lewis-acid catalyst), fries rearrangement and enantioselective glyoxalate-ene reactions.

Solubility
Soluble in water.

Notes
Hygroscopic. store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139177-62-1
Molecular Formula C3DyF9O9S3
Molecular Weight (g/mol) 609.69
MDL Number MFCD00209583
InChI Key XSVCYDUEICANRJ-UHFFFAOYSA-K
Synonym dysprosium iii trifluoromethanesulfonate, tris triflato dysprosium, dy otf 3, dysprosium iii triflate, dysprosium triflate, dysprosium trifluoromethanesulfonate, acmc-20al36, dysprosium 3+ tritriflate, dysprosium tris trifluoromethanesulfonate, dysprosium 3+ ; trifluoromethanesulfonate
PubChem CID 9938788
IUPAC Name dysprosium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Dy+3]

Specifications

Linear Formula Dy(SO3CF3)3
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 609.69
Percent Purity 98%
Chemical Name or Material Dysprosium(III) trifluoromethanesulfonate
compliance-icons
Product Identifier
  • Dysprosium (III) trifluoromethanesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Dysprosium(III) trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.