missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DZNep HCl
GREENER_CHOICE

Product Code. 18723977
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18723977 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18723977 Supplier Tocris Bioscience™ Supplier No. TB4703RMU/5
  • ONLINE EXCLUSIVE PRICE
    1250.00 €
    1181.25 € / 5mg
    Save 68.75 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

3-Deazaneplanocin A hydrochloride synthesized to Ancillary Material Grade

DZNep-RMU is an Ancillary Material Grade version of product 3-Deazaneplanocin A hydrochloride (Cat. No. 4703). Suitable for use as an ancillary reagent in the further manufacturing of cell therapies. Find out more about our Ancillary Material Grade product range.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1 S,2 R,5 R)-5-(4-Amino-1 H-imidazo[4,5-c]pyridin-1-yl)-3-(hydroxymethyl)-3-cyclopentene-1,2-diol hydrochloride
CAS 120964-45-6
Quantity 5 mg
Target EZH2 Inhibitor
Molecular Formula C12H14N4O3.HCl
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.