missing translation for 'onlineSavingsMsg'
Learn More

(E)-1-(4-(4-(2-Methylbut-2-en-1-yl)piperazin-1-yl)phenyl)ethanone Hydrochloride, TRC

Product Code. 30478115
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478115 500 mg 500mg
30465232 100 mg 100mg
30476791 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478115 Supplier Toronto Research Chemicals Supplier No. TRCM725995500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938071-78-4
Molecular Formula C17H24N2O . x(HCl)
Molecular Weight (g/mol) 308.8
Synonym Membrane Traffic Inhibitor, A5 Hydrochloride
IUPAC Name (E)-1-(4-(4-(2-methylbut-2-en-1-yl)piperazin-1-yl)phenyl)ethan-1-one hydrochloride
SMILES CC(C1=CC=C(N2CCN(C/C(C)=C/C)CC2)C=C1)=O.Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C17H24N2O.ClH/c1-4-14(2)13-18-9-11-19(12-10-18)17-7-5-16(6-8-17)15(3)20;/h4-8H,9-13H2,1-3H3;1H/b14-4+;
Chemical Name or Material (E)-1-(4-(4-(2-Methylbut-2-en-1-yl)piperazin-1-yl)phenyl)ethanone Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.