missing translation for 'onlineSavingsMsg'
Learn More

(E)-2,3,-Dehydro-cinacalcet, TRC

Product Code. 30133377
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133377 10 mg 10mg
30135272 25 mg 25mg
30009156 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133377 Supplier Toronto Research Chemicals Supplier No. TRCD22948010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 955373-56-5
Molecular Formula C22 H20 F3 N
Molecular Weight (g/mol) 355.4
Synonym 1-Naphthalenemethanamine, α-methyl-N-[(2E)-3-[3-(trifluoromethyl)phenyl]-2-propen-1-yl]-, (αR)-, (E)-N-[(1R)-1-(1-Naphthyl)ethyl]-3-[3-(trifluoromethyl)phenyl]prop-2-en-1-amine
IUPAC Name (E)-N-[(1R)-1-naphthalen-1-ylethyl]-3-[3-(trifluoromethyl)phenyl]prop-2-en-1-amine
SMILES C[C@@H](NC\C=C\c1cccc(c1)C(F)(F)F)c2cccc3ccccc23

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H20F3N/c1-16(20-13-5-10-18-9-2-3-12-21(18)20)26-14-6-8-17-7-4-11-19(15-17)22(23,24)25/h2-13,15-16,26H,14H2,1H3/b8-6+/t16-/m1/s1
Formula Weight 355.1548
Chemical Name or Material (E)-2,3,-Dehydro-cinacalcet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.