missing translation for 'onlineSavingsMsg'
Learn More

(E)-2-Decenoic acid, MedChemExpress

Product Code. 16433356
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433356 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16433356 Supplier Medchem Express Supplier No. HY13211

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress (E)-2-Decenoic acid is an interesting fatty acid isolated from royal jelly secretions of honey bees.

(E)-2-Decenoic acid is an interesting fatty acid isolated from royal jelly secretions of honey bees.

Specifications

Chemical Name or Material (E)-2-Decenoic acid
CAS 334-49-6
Color Colorless
Purity Grade Notes Research
Molecular Formula C10H18O2
Quantity 25 mg
Synonym trans-2-Decenoic acid
Solubility Information DMSO : 100 mg/mL (587.37 mM; Need ultrasonic)
SMILES OC(/C=C/CCCCCCC)=O
Molecular Weight (g/mol) 170.25
Formula Weight 170.25
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.