missing translation for 'onlineSavingsMsg'
Learn More

(E)-2-Pentenal, TRC

Product Code. 30423358
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30423358 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30423358

Brand: Toronto Research Chemicals TRCP2700055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-2-Pentenal
CAS 1576-87-0
Molecular Formula C5H8O
Quantity 5 g
Synonym 2-trans-Pentenal, trans-2-Pentenal, (2E)-2-Pentenal
InChI Formula InChI=1S/C5H8O/c1-2-3-4-5-6/h3-5H,2H2,1H3/b4-3+
SMILES CC\C=C\C=O
IUPAC Name (E)-pent-2-enal
Molecular Weight (g/mol) 84.12
Formula Weight 84.0575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.