missing translation for 'onlineSavingsMsg'
Learn More

(E)-3-(4-Amino-3,5-dimethylphenyl)acrylonitrile Hydrochloride, TRC

Product Code. 30318345
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318345 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318345 Supplier Toronto Research Chemicals Supplier No. TRCA6073401MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2E)-3-(4-Amino-3,5-dimethylphenyl)prop-2-enenitrile Hydrochloride
CAS 661489-23-2
Molecular Formula C11 H12 N2 . Cl H
Quantity 1 mg
Synonym 2-Propenenitrile, 3-(4-amino-3,5-dimethylphenyl)-, hydrochloride (1:1), (2E)-, 2-Propenenitrile, 3-(4-amino-3,5-dimethylphenyl)-, monohydrochloride, (2E)- (9CI), (E)-3-(4-Amino-3,5-dimethylphenyl)-2-propenenitrile hydrochloride
InChI Formula InChI=1S/C11H12N2.ClH/c1-8-6-10(4-3-5-12)7-9(2)11(8)13;/h3-4,6-7H,13H2,1-2H3;1H/b4-3+;
SMILES Cl.Cc1cc(\C=C\C#N)cc(C)c1N
IUPAC Name (E)-3-(4-amino-3,5-dimethylphenyl)prop-2-enenitrile;hydrochloride
Molecular Weight (g/mol) 208.69
Formula Weight 208.0767
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.