missing translation for 'onlineSavingsMsg'
Learn More

(E)-3-Acetoxy-1-propenylboronic acid pinacol ester, 97%, Thermo Scientific™

Product Code. 10482780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10482780 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10482780 Supplier Thermo Scientific Acros Supplier No. 430290010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 161395-97-7
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C11H19BO4
MDL Number MFCD03788751
Quantity 1 g
Synonym 3-acetoxy-1-propenylboronic acid pinacol ester, e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl allyl acetate, 2-3-acetoxy-1-propenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-propen-1-yl acetate, 2e-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl prop-2-en-1-yl acetate, e-3-acetoxy-1-propenylboronic acid 2,3-dimethylbutane-2,3-diyl ester
InChI Key ZXBRLGZFCXGTBA-UHFFFAOYSA-N
SMILES CC(=O)OCC=CB1OC(C)(C)C(C)(C)O1
IUPAC Name [(E)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)prop-2-enyl] acetate
Molecular Weight (g/mol) 226.08
PubChem CID 11264613
Formula Weight 226.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (E)-3-Acetoxy-1-propenylboronic acid pinacol ester
Show More Show Less
compliance-icons
Product Identifier
  • (E)-3-Acetoxy-1-propenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.