Learn More
3M™ E-A-Rfit™ Dual-Ear Validation System
Product Code. 15740068 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Product Code. Quantity Unit Size
15740068 1 Kit Each
1 options
Product Code. 15740068 Supplier 3M™ Supplier No. 7100062125
  • ONLINE EXCLUSIVE PRICE
    7815.00 €
    2994.19 € / Pack of 1
    Save 4820.81 €
    61% Off
Estimated Shipment: 25-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The 3M™ E-A-Rfit™ Dual-Ear Validation System helps determine whether workers are receiving adequate protection from their hearing protector by measuring across seven standard frequencies in under five seconds, providing a detailed report on noise attenuation performance.
  • Objective, dual-ear test, takes only seconds to administer for seven standard frequencies from 125Hz to 8000Hz
  • Includes software, speaker and impact-resistant carrying case
  • Free online training and user support
  • Requires probed test plugs, pods, or cushions
  • Dual-ear design saves time by measuring both ears simultaneously in under five seconds
  • Dual-element, dual microphone assembly is the key to the system, measuring sound levels inside and outside the hearing protector
  • Fast, clear, accurate results are obtained in just seconds and can be printed or saved electronically for future reference
  • System determines personal attenuation rating (PAR) of individual workers and helps remove measurement, fitting, and spectral uncertainty
  • Requires probed test plugs
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Approved
For Use With (Application) For automotive, food processing, general manufacturing, military, maintenance, mining, petroleum, pharmaceutical and transportation sectors
Quantity 1 Kit
Product Type Hearing Protector Fit Test Kit 393-1100
Includes Software, speaker and impact resistant carrying case