Learn More
3M™ E-A-Rfit™ Probed Test Ear Plugs
Product Code. 15789637
Change view
Click to view available options
Model:
393-2001-50
393-2002-50
393-2004-50
393-2007-50
393-2008-50
393-2010-50
Size:
Small
Universal
Product Type:
Classic™ Small
Express™
FX™
Model 1100
Push-Ins™
UltraFit™
Unit Size:
50 pairs
Pack of 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Size Product Type unitSize
15789637 393-2010-50 Universal Model 1100 Pack of 50
15739787 393-2007-50 Small Classic™ Small Pack of 50
15799777 393-2008-50 Universal Express™ Pack of 50
15729787 393-2004-50 Universal FX™ Pack of 50
15709787 393-2002-50 Universal Push-Ins™ 50 pairs
15779637 393-2001-50 Universal UltraFit™ 50 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15789637 Supplier 3M™ Supplier No. 7000002409
Only null left
Add to basket
Add to basket
Test ear plugs are modified with a probe tube for use during ear plug fit testing with the 3M™ E-A-Rfit™ Validation System
  • Science-based, quantitative testing gives accurate reports of hearing protection
  • Fast, clear, accurate results are obtained in just seconds
  • Test ear plugs are modified with a probe tube for use during ear plug fit testing with the 3M™ E-A-Rfit™ Validation System
  • 50 pair per case
  • 3M PSD products are for occupational use only
Compliance
3M™ ear plugs must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
For Use With (Application) Health & Safety Fit Testing 1100
High Visibility No
Model 393-2010-50
Packaging Type Poly bag
Pairs per Package 50 Pairs/Cs
Plug Material Polyurethane
Plug Color Orange
Corded Uncorded
Plug Shape Bullet
Product Type Model 1100
Metal Detectable No