missing translation for 'onlineSavingsMsg'
Learn More

(E)-Cinnamoyl Chloride, TRC

Product Code. 30359507
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359507 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359507 Supplier Toronto Research Chemicals Supplier No. TRCC442040250G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (E)-Cinnamoyl Chloride
CAS 17082-09-6
Molecular Formula C9H7ClO
Quantity 250 g
Synonym (2E)-3-Phenyl-2-propenoyl Chloride, (2E)-3-Phenylprop-2-enoyl Chloride, (E)-3-Phenyl-2-propenoyl chloride, (E)-Cinnamoyl Chloride, trans-3-Phenyl-2-propenoyl chloride, trans-Cinnamic Acid Chloride, trans-Cinnamoyl Chloride
InChI Formula InChI=1S/C9H7ClO/c10-9(11)7-6-8-4-2-1-3-5-8/h1-7H/b7-6+
SMILES ClC(=O)\C=C\c1ccccc1
IUPAC Name (E)-3-phenylprop-2-enoyl chloride
Molecular Weight (g/mol) 166.6
Formula Weight 166.0185
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.