missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences pASG-IBA E. coli Expression Vector with No Secretion
GREENER_CHOICE

Product Code. 15623758 Shop All IBA Lifesciences Products
Click to view available options
Vector:
pASG-IBA103
pASG-IBA105
pASG-IBA123
pASG-IBA143
pASG-IBA145
pASG-IBA162
pASG-IBA164
pASG-IBA167
pASG-IBA168
pASG-IBA23
pASG-IBA3
pASG-IBA43
pASG-IBA45
pASG-IBA5
pASG-IBA62
pASG-IBA64
pASG-IBAwt1
Unit Size:
5µg
This item is not returnable. View return policy

Product Code. 15623758

Brand: IBA Lifesciences 54168001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Regulate the expression of proteins with a vector that carries the inducible tetracycline promoter/operator. IBA Lifesciences GmbH E. coli Expression Vector with No Secretion allows for the expression of proteins in E.coli.

  • The pASG-IBA vectors are designed for expression of target proteins without affinity tag in E. coli.
  • The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, and the inducible tetracycline promoter/operator for the regulated expression of proteins.
  • The vectors can be combined with any E. coli strain since the tet-promoter works independently of the genetic background of E. coli.
  • Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity.
Please note that cloning into pASG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pASG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.

Specifications

Inducing Agent Inducible with anhydrotetracycline
Marker Gene Ampicillin
Promoter Tet
Product Type Vector
Content And Storage Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
Species E.coli
Protein Tag Twin-Strep-tag-FLAG-tag
Concentration 250 ng/μL
Primer Forward
Quantity 5 μg
Vector pASG-IBA168
For Use With (Application) Vector for recombinant expression in E. coli
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.