missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences pASG-IBA E. coli Expression Vector with Secretion
Regulate the expression of proteins with a vector that carries the inducible tetracycline promoter/operator. E. coli Expression Vector with Secretion allows for the expression of proteins in E. coli.
266.00 € - 286.00 €
Specifications
Inducing Agent | Inducible with anhydrotetracycline |
---|---|
Marker Gene | Ampicillin |
Promoter | Tet |
Product Type | Vector |
Content And Storage | Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage |
Product Code | Brand | Protein Tag | Vector | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Protein Tag | Vector | Price | Quantity & Availability | ||||
15613748
|
IBA Lifesciences
5-4102-001 |
Twin-Strep-tag | pASG-IBA102 |
266.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15633748
|
IBA Lifesciences
5-4104-001 |
Twin-Strep-tag | pASG-IBA104 |
266.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15643728
|
IBA Lifesciences
5-4001-001 |
Untagged | pASG-IBAwt2 |
266.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15643738
|
IBA Lifesciences
5-4044-001 |
His Tag (6x), Strep-tag II | pASG-IBA44 |
266.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15653728
|
IBA Lifesciences
5-4002-001 |
Strep-tag II | pASG-IBA2 |
266.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15673728
|
IBA Lifesciences
5-4004-001 |
Strep-tag II | pASG-IBA4 |
266.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15673748
|
IBA Lifesciences
5-4144-001 |
His Tag (6x), Twin-Strep-tag | pASG-IBA144 |
286.00 €
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Regulate the expression of proteins with a vector that carries the inducible tetracycline promoter/operator. IBA Lifesciences GmbH E. coli Expression Vector with Secretion allows for the expression of proteins in E.coli.Specifications
Inducible with anhydrotetracycline | |
Tet | |
Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage | |
ompA for periplasmic secretion |
Ampicillin | |
Vector | |
5 μg |