missing translation for 'onlineSavingsMsg'
Learn More

E.coli Genomic DNA

Product Code. 15649898 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15649898 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15649898 Supplier Thermo Scientific Supplier No. J14380.MA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 9007-49-2 | C15H31N3O13P2

 

  • Single chromosome
  • Genome length: 4600000 bp
  • Fragmented to some degree during purification yet it is characterized as high molecular weight DNA
TRUSTED_SUSTAINABILITY

Specifications

CAS 9007-49-2
Molecular Formula C15H31N3O13P2
MDL Number MFCD00130921
InChI Key AWBASQCACWFTGD-UHFFFAOYNA-N
SMILES *
IUPAC Name Deoxyribonucleic acid
Molecular Weight (g/mol) 0.00
Product Type Genomic DNA
Quantity 10 mg
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.