missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ CloneWell™ II Agarose Gels with SYBR Safe, 0.8%
GREENER_CHOICE

Product Code. 15796548
Change view
Click to view available options
Quantity:
10 gels
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15796548 10 gels Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15796548 Supplier Invitrogen™ Supplier No. G661818
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Includes

Includes 10 gels, and 1 vial 10X Sample Loading Buffer

Faster and more effective DNA fragment isolation method for successful cloning workflow. Recover your DNA in 3 easy steps.

E-Gel CloneWell II Agarose Gels with SYBR Safe enable faster and more effective DNA fragment isolation for cloning workflows. The use of in-gel SYBR Safe stain and a blue-light transilluminator minimizes DNA damage and improves cloning efficiency compared to conventional methods. The E-Gel CloneWell II agarose gels are designed for use with the E-Gel Power Snap Electrophoresis Device (Cat. No. G8100).


E-Gel CloneWell II agarose gels enable you to:

  • Save time—no additional gel purification steps required after electrophoresis
  • Simplify DNA recovery—retrieve purified DNA directly from the recovery well with a pipette
  • Improve cloning results—minimize DNA damage and get more colony forming units for any cloning method

Fast DNA sample isolation in three steps
DNA fragment isolation with E-Gel CloneWell II precast gels is a simple 3-step procedure that eliminates conventional gel purification. To extract the DNA band of interest, load the sample into the loading wells, run the gel until the band reaches the recovery wells, and retrieve the sample. The covered sample is ready for cloning and does not require re-purification. Multiple bands from the one loading well can be collected during the same gel run.

DNA extracted from CloneWell II gels is compatible with common cloning methods such as Anza restriction cloning, TOPO cloning, and Gateway cloning.

Real-time control of the sample
SYBR Safe DNA Gel Stain is incorporated into the gel itself, allowing you to visually monitor DNA band migration progress on the blue-light transilluminator of the E-Gel Power Snap Electrophoresis Device. The gel has reference marks for better control of the sample prior to the sample recovery step. If the target DNA passes the recovery well, the Reverse gel protocol allows for re-capture of the DNA band.

Improved cloning efficiencies
Even a short exposure of your DNA sample to UV light during visualization may lead to DNA damage and reduced cloning efficiencies. Using E-Gel CloneWell II gels together with the blue-light transilluminator minimizes UV damage and improves cloning efficiency when compared to conventional methods.

Safer alternative to ethidium bromide
The E-Gel CloneWell II agarose gel is pre-stained with SYBR Safe DNA stain that minimizes environmental impact and is safer than the ethidium bromide used in conventional preparative gel electrophoresis.

TRUSTED_SUSTAINABILITY

Specifications

Compatible Ladders E-Gel™ 1 Kb Plus Express DNA Ladder
Content And Storage Store at room temperature.
Gel Percentage 0.8%
Gel Type E-Gel
Label or Dye SYBR Gold
Wells 7-well
Quantity 10 gels
Shipping Condition Room Temperature
Product Type Agarose Gel

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.