missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, SYBR Safe 1%

Product Code. 17741478
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17741478 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17741478 Supplier Invitrogen™ Supplier No. G8121ST
Limited-Time Offers on Selected Molecular Biology Essentials
Use promo code 27945
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Provides everything needed to quickly start using precast 1% E-Gel agarose gels with SYBR Safe on the device.

The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, SYBR Safe 1%, provides everything you need to quickly start using precast 1% E-Gel agarose gels with SYBR Safe on the device.

The kit includes:

  • 10 E-Gel agarose gels with SYBR Safe, 1% (Cat. No. A42100)
  • E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel 1 Kb Plus DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488090)
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 1 electrophoresis device
• 10 E-Gel agarose gels with SYBR Safe, 1%
• 1 vial E-Gel 1 kb Plus DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses

Store at room temperature.

Product Line E-Gel
Type Electrophoresis System Starter Kit
Quantity 1 Kit
Shipping Condition Room Temperature
Max. Voltage 240 VAC

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.