Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, SYBR Safe 2%
Description
Includes
E-Gel Power Snap Electrophoresis device,E-Gel Power Snap Camera,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.
The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, SYBR Safe 2%, provides everything you need to quickly start using precast 2% E-Gel agarose gels with SYBR Safe on the system for a streamlined gel electrophoresis and gel imaging workflow.
The kit includes:
- 18 E-Gel agarose gels with SYBR Safe, 2% (Cat. No. G521802)
- E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
- E-Gel 50 bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
Warranty
1 year warranty
Specifications
Specifications
| Content And Storage | • 1 electrophoresis device • 1 camera device • 18 E-Gel SYBR Safe 2% gels • 1 vial E-Gel 50 bp DNA Ladder • 1 power adapter • 1 pair of viewing glasses • 1 E-Gel Go! Adapter Store at room temperature. |
| Type | Electrophoresis System Starter Kit |
| Shipping Condition | Room Temperature |
| Max. Voltage | 240 V AC (Power Supply), 48 V DC (Electrophoresis Device) |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.