missing translation for 'onlineSavingsMsg'
Learn More

(E)-Naftifine, TRC

Product Code. 30477886
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477886 10 mg 10mg
30475006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477886 Supplier Toronto Research Chemicals Supplier No. TRCN21331010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65472-88-0
Molecular Formula C21 H21 N
Molecular Weight (g/mol) 287.4
Synonym 1-Naphthalenemethanamine, N-methyl-N-[(2E)-3-phenyl-2-propen-1-yl]-, 1-Naphthalenemethanamine, N-methyl-N-(3-phenyl-2-propenyl)-, (E)-, 1-Naphthalenemethanamine, N-methyl-N-[(2E)-3-phenyl-2-propenyl]- (9CI), N-Methyl-N-[(2E)-3-phenyl-2-propen-1-yl]-1-naphthalenemethanamine, Nafetifine, Naftifine, Naftifungin
IUPAC Name (E)-N-methyl-N-(naphthalen-1-ylmethyl)-3-phenylprop-2-en-1-amine
SMILES CN(C\C=C\c1ccccc1)Cc2cccc3ccccc23

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H21N/c1-22(16-8-11-18-9-3-2-4-10-18)17-20-14-7-13-19-12-5-6-15-21(19)20/h2-15H,16-17H2,1H3/b11-8+
Formula Weight 287.1674
Percent Purity >95%
Chemical Name or Material (E)-Naftifine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.