missing translation for 'onlineSavingsMsg'
Learn More

(E)-Olopatadine Hydrochloride, TRC

Product Code. 30005068
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005068 1 mg 1mg
30005940 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005068 Supplier Toronto Research Chemicals Supplier No. TRCO5750051MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 949141-22-4
Molecular Formula C21 H23 N O3 . Cl H
Molecular Weight (g/mol) 373.87
Synonym (11E)-11-[3-(Dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic Acid Hydrochloride (1:1), (E)-Olopatadine Hydrochloride
IUPAC Name 2-[(11E)-11-[3-(dimethylamino)propylidene]-6H-benzo[c][1]benzoxepin-2-yl]acetic acid;hydrochloride
SMILES Cl.CN(C)CC\C=C\1/c2ccccc2COc3ccc(CC(=O)O)cc13

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H23NO3.ClH/c1-22(2)11-5-8-18-17-7-4-3-6-16(17)14-25-20-10-9-15(12-19(18)20)13-21(23)24;/h3-4,6-10,12H,5,11,13-14H2,1-2H3,(H,23,24);1H/b18-8+;
Formula Weight 373.1445
Percent Purity >95%
Chemical Name or Material (E)-Olopatadine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.