missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-PAGE™ Midi Protein Gels, 3.7 mm

Product Code. 10654583
Change view
Click to view available options
Wells:
48-well
96-well
Gel Percentage:
6%
8%
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wells Gel Percentage Unit Size
10654583 96-well 6% Each
10206172 48-well 8% Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10654583 Supplier Invitrogen™ Supplier No. EP09606
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

8 E-PAGE™ 96 gels, 4.5mL E-PAGE™ Loading Buffer 1 (4X), 1 Butterfly Opener, 1 E-PAGE™ Blotting Pad

E-PAGE™ Gels are self-contained, pre-cast gels that include a gel matrix and electrodes packaged inside a disposable cassette.

E-PAGE Gels are self-contained, pre-cast gels that include a gel matrix and electrodes packaged inside a disposable cassette. Protein separation on an E-PAGE Gel requires an EBase Integrated Device, a compact and automated platform designed for medium- and high-throughput electrophoresis.

Each E-PAGE 8% 48-well gel contains 48 sample lanes and 4 marker lanes with a 3.2-cm separation length. E-PAGE 48-well gels can be loaded with a 12- or 8-channel pipettor or 4- or 8-tip robotic system (Figure 1).

Each E-PAGE 6% 96-well gel contains 96 sample lanes and 8 marker lanes with a 1.6-cm separation length. The well openings of the cassette are compatible with the standard 96-well plate format and can be conveniently loaded with a multichannel pipettor or 8-, 12-, or 96-tip liquid-handling robotic device (Figure 2).

Both E-PAGE 48 and 96 cassettes can be opened to remove the gel for downstream staining or blotting (Figures 3 and 4). In addition, each cassette is labeled with an individual barcode to facilitate identification of the gel using commercial barcode readers.

More about the E-PAGE precast gel system
The Invitrogen E-PAGE precast gel system is designed for fast, mess-free medium- and high-throughput protein electrophoresis. It consists of the E-PAGE precast gels, E-Base Integrated devices for running the gels, and free E-Editor 2.0 software for image analysis.

The all-in-one E-Gel Power Snap Plus Electrophoresis System is designed for fast and convenient high-throughput gel electrophoresis. With dry precast E-PAGE gel technology, you can run protein samples in as little as 14 minutes. The device arrives with pre-programmed protocols for both E-PAGE gel types.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Gel Percentage 6%
Gel Size Midi
Gel Thickness 3.7 mm
Gel Type E-PAGE
Sample Loading Volume Up to 20 μL
Separation Range 10 to 300 kDa
Separation Type Denaturing
Wells 96-well
Mode of Separation Molecular Weight
Quantity 8 gels (1 kit)
Shipping Condition Room Temperature
Storage Requirements Store at room temperature, do not allow the temperature to drop below 4°C or rise above 40°C
Length (Metric) 8 cm
Width (Metric) 13 cm
Product Line E-PAGE
For Use With (Equipment) E-BASE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.