Learn More
Thermo Scientific™ E1-ClipTip™ Electronic Pipette Charging Stand
Product Code. 15296283
Change view
Click to view available options
For Use With:
1 Pipette
3 Pipettes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
15296283 1 Pipette Each
15345961 3 Pipettes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15296283 Supplier Thermo Scientific™ Supplier No. 9420500
  • ONLINE EXCLUSIVE PRICE
    414.00 €
    301.01 € / Pack of 1
    Save 112.99 €
    27% Off
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Conveniently charge and store one or three pipettes with these custom charging stands.
Conveniently charge and store one or three E1-ClipTip pipettes with the Thermo Scientific™ E1-ClipTip™ Charging Stands.
Warranty: 1 year
TRUSTED_SUSTAINABILITY
Specifications
For Use With 1 Pipette
Type Charging Stand
For Use With (Equipment) E1-ClipTip Electronic Single Channel Pipette
Includes Universal power supply