Learn More
Berkshire Corporation EasyClean™ 360 Isolator Knitted Polyester Padded Covers
BIOPHARMA
Product Code. 17523286 Shop All Berkshire Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 200
Product Code. Color Unit Size
17523286 White Pack of 200
1 options
Product Code. 17523286 Supplier Berkshire Corporation Supplier No. EC360CHP40
  • ONLINE EXCLUSIVE PRICE
    1522.50 €
    790.74 € / Pack of 200
    Save 731.76 €
    48% Off
Estimated Shipment: 15-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The EasyClean™ 360 Isolator Cleaning System includes a light-weight, maneuverable cleaning tool along with knitted polyester covers.
  • Made from Berkshire's Choice™ SuperSorb 100% continuous filament knitted polyester material
  • Laundered and packaged in an ISO Class 4 cleanroom for superior fiber, particle and residue performance
  • Foam-free cushioned design ensures even surface coverage with reduced residues and contaminants
  • Easy installation and secure fit on the mop head
  • Recommended for use with the EasyClean™ 360 Knitted Polyester Elastic Covers
  • Compatible for use in ISO Class 3/EU Grade A environments after sterilizing
Specifications
Product Type Mop Cover
Color Code White
Color White
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Single-use
Color-coded No
Material Polyester