missing translation for 'onlineSavingsMsg'
Learn More

SCHOTT AG Lighting and Imaging™ EasyLED Spotlight Plus

Product Code. 15489763 Shop All Schott Products
Change view
Click to view available options
Type:
Double Spot Illumination
Single Spot Illumination
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
15489763 Single Spot Illumination Each
15499763 Double Spot Illumination Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489763 Supplier SCHOTT AG Lighting and Imaging™ Supplier No. 600101
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The EasyLED Spotlight Plus is an ergonomic and compact LED spot illumination system for stereo microscopy and macroscopy. Power supply included.

  • Advanced spot illumination.
  • Can be mounted to any microscope stand or pillar.
  • Dimming control is ergonomically placed to the focus control of the microscope
  • Robust metal light head.
  • Equipped with special cooling lamellae that prevent the illuminated object from heating up under the microscope.
  • Provide an attractive alternative to conventional cold light sources with fiber optics.
  • Single or double lighting head.
  • Adapters for all stands plus stand-alone version available.
  • Mounting bracket need to be ordered separately.
  • Integrated controller.
  • Extra contrast with alternating light.
  • Wide range power supply.
  • International clip plug system.
  • Spot Light Plus Double can be controlled individually.
  • Aviation proofed-flex and stay technology.

Specifications

Type Single Spot Illumination
Bulb Type LED
Certifications/Compliance CE
Description Single light head
Dimensions (L x W x H) 61 x 114 x 107 mm Control Unit
For Use With (Application) Stereo Microscopy Illumination
For Use With (Equipment) Mounting Bracket
Frequency 50/60 Hz
Includes Power supply
Lamp Life 50,000 Hours
Length (Metric) 500 mm (incl. light head)
Lighting Type High Power LED illumination
Range (Lux) 130 lm
Voltage 100/240 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.