missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EasyPep™ MS Sample Prep Kits

Product Code. 16874571 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Lysis Buffer
MS Sample Prep Kit
Maxi Sample Prep Kit
Micro MS Sample Prep Kit
Mini MS Sample Prep Kit
Peptide Clean-up Plate
Unit Size:
122g
20 tests
8 reactions
96 reactions
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
16874571 Lysis Buffer 122g
15947032 Mini MS Sample Prep Kit 20 tests
17845047 Micro MS Sample Prep Kit 96 reactions
17175772 MS Sample Prep Kit 96 reactions
17185772 Maxi Sample Prep Kit 8 reactions
17895257 Peptide Clean-up Plate 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 16874571 Supplier Thermo Scientific™ Supplier No. A45735

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

Generate high quality proteomic samples for MS analysis in less than four hours using optimized EasyPep MS Sample Prep kits.

EasyPep Mini MS Sample Prep Kits enable efficient and reproducible processing of cultured mammalian cells and tissues for proteomic MS analysis. The kit contains pre-formulated buffers, MS-grade enzyme mix, peptide clean-up columns, and an optimized protocol to generate MS-compatible peptide samples in less than three hours. Kits are optimized to process protein samples from 10–100 μg with a high yield of MS-ready peptides.

Complete—includes pre-formulated reagents for lysis through digestion, peptide clean-up columns, and an optimized protocol for up to 20 samples
Optimized—streamlined protocol and reagents minimize the number of steps and time time needed to process samples
Flexible—reagents and protocol have been verified using cells, plasma, and tissue samples for 10 to 100 μg samples
Time-saving—sample processing time reduced from more than one day to less than three hours
Compatible—final preparation is ready for direct MS analysis and other downstream applications, including TMT labeling

The sample preparation of peptides for MS analysis is complex, with numerous steps and varied protocols, resulting in sample variability in both quality and quantity. This kit has been designed to improve reproducibility while saving hands-on and processing time. The number of steps and processing time have been reduced through the addition of Universal Nuclease to reduce viscosity from nucleic acids (replacing sonication), a rapid 'one pot' reduction/alkylation solution for cysteine modification (carbamidomethylation, + 57.02), and a trypsin/Lys-C protease mix for more complete digestion. In addition, the kit includes Peptide Clean-up columns and buffers to prepare detergent-free peptide samples for MS analysis. The optimized reagents and protocol produce high-quality peptides that are compatible with TMT labeling and other downstream applications, or are ready for MS analysis.

TRUSTED_SUSTAINABILITY

Specifications

Description Lysis Buffer
For Use With (Equipment) Mass Spectrometer
Product Type Lysis Buffer
Product Line EasyPep
Content And Storage Store at 2°C to 8°C
Label or Dye Unlabeled
Shipping Condition Wet Ice
Starting Material Protein Samples
Final Product Type Protein
Detection Method Mass Spectrometry
Workflow Step Cell Lysis
Quantity 100 mL
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.