missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ EasySampler™ Sampler For Solids

Product Code. 10648534 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
60 mL
200 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10648534 200 mL Each
15529650 60 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10648534 Supplier Buerkle™ Supplier No. 53601000
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Quick and easy sampling of free-flowing bulk materials

The EasySampler offers an innovative solution for fast sampling and emptying of free-flowing bulk materials by allowing the sample to be emptied quickly and directly into the collection container. By pressing down on the handle, the sampler tip opens and the sample can be emptied without turning or dismantling the sampler.

The smooth, rear-edge-free design ensures complete emptying and prevents the build-up of residual material, allowing fast and repeated sampling.

In addition, the sampler can be easily disassembled for thorough cleaning. The use of high quality stainless steel 316L (1.4404) guarantees durability, reliability and purity during sampling.

The EasySampler is specially designed for free flowing or self-flowing materials and greatly simplifies sampling.

  • Innovative design for quick and easy sampling
  • Empty directly into collection container
  • High quality stainless steel for durability and reliability
  • Easily disassembled for thorough cleaning
  • Optimised for free flowing or self flowing bulk solids
TRUSTED_SUSTAINABILITY

Specifications

Product Type Sample Lance
Material Stainless Steel (V4A)
Autoclavable Autoclavable
Capacity (Metric) 200 mL
Length (Metric) 1045 mm
Length (Metric) Bowl 500 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.