missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ HLA Chimerism Whole Blood CD8 Positive Selection Kit

Product Code. 17197801
Encompass
Change view
Click to view available options
Quantity:
60 mL whole blood
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17197801 60 mL whole blood 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17197801 Supplier STEMCELL Technologies Supplier No. 17889

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Column-free immunomagnetic positive selection of human CD8+ cells for HLA analysis

  • The EasySep™ HLA Chimerism Whole Blood CD8 Positive Selection Kit is designed to isolate CD8+ cells from fresh whole blood or buffy coat by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD8 and dextran-coated magnetic particles
  • The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • The CD8 antigen is expressed on cytotoxic T cells and weakly on a subset of NK cells
  • This product replaces the EasySep™ Human Whole Blood CD8 Positive Selection Kit (Catalog #18083) for even faster cell isolations

Specifications

Cell Type T Cells, CD8+
For Use With (Application) Cell Isolation
Quantity 60 mL whole blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.