missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Essential Human T-Cell Phenotyping Panel

Product Code. 16301994
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16301994 100 reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16301994 Supplier Invitrogen™ Supplier No. A42806
  • ONLINE EXCLUSIVE PRICE
    1182.00 €
    839.22 € / 100 reactions
    Save 342.78 €
    29% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Invitrogen™ eBioscience™ Essential Human T-Cell Phenotyping Panel is designed for flow cytometry-based identity testing of T-cells

Essential Human T-Cell Phenotyping Panel everages high quality eBioscience™ antibodies and a ready-to-use protocol for experimental setup, gating strategy, and analysis.

Benefits include:

  • Proven performance―antibodies designed for flow cytometry
  • Highly referenced antibodies―eBioscience™ portfolio contains trusted fluorochrome-conjugated antibodies for immunology and oncology research
  • Turnkey kit and protocol―contains standardized gating recommendations needed to perform flow cytometry characterization
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage CD3, CD4, CD8, CD62L, and CCR7 antibodies, store at 2°C to 8°C
Format Kit
Quantity 100 reactions
Detection Method Fluorescence
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Line eBioscience
Product Type Phenotyping Panel

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.