missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Essential Human Treg Phenotyping Panel & Staining Buffers

Product Code. 17134729
Change view
Click to view available options
Quantity:
100 reactions
Unit Size:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17134729 100 reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17134729 Supplier Invitrogen™ Supplier No. A42924
  • ONLINE EXCLUSIVE PRICE
    1246.00 €
    884.66 € / 100 reactions
    Save 361.34 €
    29% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Invitrogen™ eBioscience™ Essential Human Treg Phenotyping Panel and Staining Buffers are designed for flow cytometry-based identity testing of regulatory T-cells (Tregs).

Phenotyping Panel and Staining Buffers leverages high quality eBioscience™ antibodies and fixation/permeabilization reagents (for the intracellular marker FOXP3) and includes a ready-to-use protocol for experimental set-up, gating strategy, and analysis.

Benefits include:

  • Proven performance―antibodies and supporting reagents designed for flow cytometry
  • Highly referenced antibodies―includes one of the most referenced clones for FOXP3
  • Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage
  • CD4, CD25, CD127, and FOXP3 antibodies, store at 2–8°C
  • FOXP2/TF Staining Buffer set, store at 2–8°C
Format Kit
Quantity 100 reactions
Detection Method Fluorescence
Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Line eBioscience
Product Type Phenotyping Panel and Staining Buffer

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.