missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Immune Response T Cell Receptor Signaling Kit

Product Code. 17787473
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17787473 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17787473 Supplier Invitrogen™ Supplier No. A53422

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Kit is comprised of positive control stimulation reagents, buffers, viability dye, antibodies, and compensation beads needed to stimulate and detect phosphorylation of CD247 (CD3 zeta) on tyrosine 142 (Tyr142) in human T cells by flow cytometry.

  • Kit is comprised of positive control stimulation reagents, buffers, viability dye, antibodies, and compensation beads
  • Use to stimulate and detect phosphorylation of CD247 (CD3 zeta) on tyrosine 142 (Tyr142) in human T cells by flow cytometry
  • Unique spectral fingerprint is designed to provide the brightest signal for flow cytometry experiments
  • Complete kit packaged in single-use vials for ease of use, and to ensure stability performance over time
  • Extensive validation and protocol optimization for consistent results
  • Stimulation, fixation, permeabilization, and staining are performed in 96-well, round-bottom plates
  • Alternatively, cells may be cultured in bulk and transferred to 12 x 75 mm test tubes
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Antibodies, compensation beads, viability stain, buffers
    Store at 4°C and –20°C.
    For Use With (Application) T Cell Receptor Signaling Analysis and Research
    For Use With (Equipment) Flow Cytometry
    Quantity 1 Kit
    Regulatory Status RUO
    Shipping Condition On Ice
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.