missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ EC 10 Universal Alkaline Cleaning Concentrate

Product Code. 10406851 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10406851 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10406851
  • ONLINE EXCLUSIVE PRICE
    20.55 €
    20.00 € / 1 L
    Save 0.55 €
    2% Off
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Suitable for thorough ultrasonic cleaning of medical and detal instruments and laboratory equipment made of stainless steel, glass, plastics and ceramics.

  • Removes blood, saliva, proteins and bone residues as well as grease, oil, grinding and polishing plates
  • Not suitable for aluminum, zinc or light metal alloys and for plastics that are susceptible to stress cracking
  • Store in closed original container at a temperature between 5° and 30°C, protected from heat and direct solar radiation
  • Shelf life of 5 years from date of production

Specifications

Product Type Ultrasonic Cleaning Solution
Composition Alkaline
Content And Storage Between 5°C and 30°C, original container, away from heat and direct solar radiation
For Use With (Application) Ultrasonic cleaning of medical and dental instruments
Volume (Metric) 1 L
compliance-icons
Product Identifier
  • bioson 10
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: disodum tetraboratedecahydrate, sodiumhydroxide, n-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.