missing translation for 'onlineSavingsMsg'
Learn More

Elma™ EC 210 Cleaner

Product Code. 11741369
Change view
Click to view available options
Volume (Metric):
25 L
Unit Size:
25 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11741369 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11741369 Supplier Elma™ Supplier No. 5809800000
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ultrasonic cleaning concentrate for metal and plastic surfaces. Elma™ EC 210 Cleaner has a 3-year shelf life.

  • Suitable for surfaces made of steel, nonferrous heavy metals, precious metals, light metals, nickel, chrome, zinc, and German silver (also combinations of different materials); it is also suitable for plastics that are resistant against stress cracking.
  • Check any Mg, MgZnAl alloys and plastics for compatibility before application.
  • Removes aqueous cooling lubricants, grease and oil abrasive, grinding and polishing pastes.
  • For the cleaning of polishing media containing animal greases, check the suitability of the cleaner before application.

Specifications

Form Liquid
pH 8.9
Appearance Light Beige
Boiling Point >100°C
Composition Anionic surfactants 5 to 15%, non-ionic surfactants 15 to 30%, perfumes
Concentration Ultrasonic bath 3 to 5%
Container Type Can
Content And Storage -5°C to 30°C
Volume (Metric) 25 L
compliance-icons
Product Identifier
  • elma clean 210 (EC 210)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.