Learn More
Macherey Nagel EC HPLC-Column EC 125/2NUCLEODUR PFP, 3 µm length: 125 mm,
Brand:  Macherey Nagel 760447.20
EC HPLC-Column EC 125/2NUCLEODUR PFP, 3 µm length: 125 mm,
Product Code. 12768782
  • ONLINE EXCLUSIVE PRICE
    815.00 €
    780.00 € / Each
    Save 35.00 €
    4% Off