Learn More
Trajan Scientific and Medical™ EC Science Silicon Septa
Product Code. 15248623
Change view
Click to view available options
Diameter (Metric) Inner:
9.5 mm
11 mm
11.5 mm
12.5 mm
Unit Size:
Pack of 24
Pack of 25
Pack of 50
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
15248623 12.5 mm Pack of 24
15258623 11 mm Pack of 50
15218623 9.5 mm Pack of 25
15228623 11.5 mm Pack of 25
15238623 - Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15248623 Supplier Trajan Scientific and Medical™ Supplier No. 041906
Add to basket
Add to basket
Designed for long lifetime and low injection port adhesion.
  • Combines long injection lifetime, low bleed and low injection port adhesion
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications
Type EC
Diameter (Metric) Inner 12.5 mm
Quantity 24/Pk.