missing translation for 'onlineSavingsMsg'
Learn More

Sharpsguard™ Eco 11.5 Waste Container

Product Code. 17764854
Change view
Click to view available options
Color:
Orange
Yellow
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17764854 Yellow Each
17754854 Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17764854 Supplier Sharpsguard™ Supplier No. DD476YLR
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

The new Sharpsguard™ eco containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have not been contaminated with medicines.

  • Sharpsguard™ eco 11.5 is a multi-purpose container featuring a robust pail handle for easy transportation. A wider aperture allows easy insertion of larger items.
  • This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • It is designed to fit safely and securely into our POUDS™ mobile holders and bracket range.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-0

Specifications

Capacity (Metric) 9.27 L
Diameter (Metric) Outer 275 mm
Color Yellow
Height (Metric) 249 mm
Quantity 1 Ea.
Width (Metric) 622 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.