missing translation for 'onlineSavingsMsg'
Learn More

PCRmax Eco 48 Real-Time qPCR System 48 Well Block

Product Code. 17453203
Change view
Click to view available options
Content And Storage:
Plate Dock, 48-well Plates and Seals
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Content And Storage Unit Size
17453203 Plate Dock, 48-well Plates and Seals Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17453203 Supplier PCRmax Supplier No. 9394700
  • ONLINE EXCLUSIVE PRICE
    19227.44 €
    18250.23 € / Pack of 1
    Save 977.21 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The PCRmax Eco 48 utilizes patented block and optical technologies to deliver extreme speed, sensitivity and data quality.

The PCRmax Eco48 real time PCR system is a high specification, economically priced real time thermal cycler that accommodates a unique 48-well polypropylene PCR plate utilizing the same geometry as standard 384-well plates, but only 1/8 of the size. This enables users to dramatically reduce the qPCR reagent volumes compared to traditional 96-well instruments, while still producing a strong fluorescence signal. Minimizing the plate size also significantly improves thermal uniformity. A minimum volume of 5 μL is validated, cutting reagent costs in half and making more efficient use of expensive and hard to acquire template DNA samples.

The easy-to-use Eco48 system comes with two distinct softwares. Eco Control drives the units setup and experimental runs while Eco Study is the analysis package. Eco48 software is open license and MIQE compliant, so you can install it on as many PCs as you would like.

  • MIQE compliant open license software gives confidence when publishing data
  • Eco48 allows for simple multiplex analysis with up to four dyes at once
  • Fast cycling enables several experiments per day, all at an economical price
  • Eco48 is open platform and can utilize any chemistry, dye or PCR reagent
  • Eco 48 system monitors data collection in real time, allowing researchers to access run viability immediately
  • User-friendly interface allows researchers to easily view amplification plot, melt curve analysis, and the analyzed results, including Cq values, PCR efficiency, R2 and Y-intercept

Specifications

Block Format Peltier-based
Thermal Accuracy ±0.25°C
Temperature Range (Metric) 30 to 100°C
Content And Storage Plate Dock, 48-well Plates and Seals
Thermal Uniformity ±0.1°C
Reaction Speed 40 min. Standard Run Time
Voltage 100/240 V
Display Type PC or Laptop Interface
Description Proprietary Hollow Silver/Gold Block
Max. Heating Rate (Thermal Block) 5.5°C/s
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.