missing translation for 'onlineSavingsMsg'
Learn More

Sharpsguard™ Eco Cyto 2.5 Waste Container
GREENER_CHOICE

Product Code. 17714874
Change view
Click to view available options
Capacity (Metric):
2.21 L
Unit Size:
Pack of 48
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17714874 2.21 L Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17714874 Supplier Sharpsguard™ Supplier No. DD672R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The new Sharpsguard™ eco cyto containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have been contaminated with cytotoxic or cytostatic medicines.

  • Sharpsguard™ eco cyto 2.5 is an ideal general-purpose sharps container for cytotoxic and cytostatic waste, it provides maximum capacity in the minimum working space.
  • This container is supplied with the base and lid separately and contains an absorbent pad. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • It is designed to fit safely and securely into our POUDS™ tray range.
  • This container is for disposal by incineration or other authorised process and is designed for the consignment of waste categorised by EWC codes: 18-01-01, 18-01-03, 18-01-08, 18-01-09, 18-02-01, 18-02-02, 18-02-0, 18-02-08.

Eco Cyto, Waste Container, cytotoxic, cytostatic, sharpsguard

Specifications

Capacity (Metric) 2.21 L
Closure Color Purple
Diameter (Metric) Outer 176 mm
Color Yellow
Height (Metric) 166 mm
Model Eco Cyto 2.5
Quantity 48/Pk.
Weight (Metric) 232 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.