Learn More
Skytec™ Eco Nickel Gloves
GREENER_CHOICE
Product Code. 30274625
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
Product Code. 30274625
Brand: Skytec™ SKG00053BL
Only null left
Add to basket
Add to basket
Lightweight general handling glove with PU coating and recycled polyester.
  • General Handling Glove with PU Coating
  • Ideal for detailed assembly
  • 1 pair = 2 x 500 mL (PET) plastic bottles
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Abrasion 4,Puncture 2, EN 388:2016 + A1:2018 3131X, EN 21420:2020
Construction Seamless Knit
Coating Material Polyurethane
Color Black
Exterior Finish Rough
Hand Ambidextrous
Latex Free Yes
Material Recycled Polyester
Product Type General Purpose Gloves
Size 2X-Large
Length (Metric) 270 mm