missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ ECO16 Thermoreactor for COD Analysis

Product Code. 11761953 Shop All Velp Products
Change view
Click to view available options
Range:
1 to 199 min. or continuous Timer
Unit Size:
Each
Product Code. Range unitSize
11761953 1 to 199 min. or continuous Timer Each
1 options
This item is not returnable. View return policy

Product Code. 11761953

Brand: Velp Scientifica™ F10100126

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Velp Scientifica™ ECO16 Thermoreactor for COD Analysis with settable temperature (up to 165°C) and operating time (up to 999 minutes).

  • Electronic temperature control
  • Temperature regulation from ambient to 165°C
  • Analysis time can be set from 1 to 999 minutes or continuous
  • Processes up to 14 x Ø 16 mm mL test tubes
  • Plus 2 x Ø 22 mm test tubes simultaneousl
  • LCD countdown display shows the temperature and remaining time
  • COD analysis in only 30 minutes at 160°C
  • 16 mm diameter tubes minimize reagent consumption
  • Automatic shut-down at run end
  • Acoustic signals, e.g., at shut-down
  • Anti-splash cover protects the user

Specifications

Certifications/Compliance CE, CB Certificate, cNEMKOus
For Use With (Application) Chemical oxygen demand analysis
No. of Tests 16
Range 1 to 199 min. or continuous Timer
Test Method COD
Type 230 V / 50-60 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.