missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Electronic temperature control
- Temperature regulation from ambient to 200°C
- Analysis time can be set from 1 to 999 minutes or continuous
- Processes up to 6 x 200 mL test tubes, Ø 42 mm or 6 x 22 mm test tubes or 18 Ø 16 mm test tubes
- LCD countdown display shows the temperature and remaining time
- COD analysis in only 30 minutes at 160°C
- 16 mm diameter tubes minimize reagent consumption
- LCD display for immediate understanding
- Automatic shut-down at run end
- Acoustic signals, e.g., at shut-down
- Antisplash bells protect the user and other sample
Specifications
Specifications
| Description | Chemical Oxygen Demand Thermoreactor |
| For Use With (Application) | Analysis of water |
| Range | 1 to 199 min. or continuous Timer |
| Test Method | COD |
| Type | 230 V / 50-60 Hz |
| Dimensions (L x W x H) | 198 x 132 x 319 mm |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?