missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientific ECODryBlock / Dry Block Heater

Product Code. 31703667 Shop All Velp Products
Change view
Click to view available options
Temperature Range (Metric):
5°C to 165°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) unitSize
31703667 5°C to 165°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31703667 Supplier Velp Scientific Supplier No. SA101A0700
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Modular, digital, and precise dry block heater designed for sample preparation, incubation, DNA denaturation, enzymatic reactions, and culture media tests.

Velp Scientifica ECODryBlock is a modern, versatile dry block heater designed for precise temperature control and flexible laboratory workflows.

Key Features

  • Modern, intuitive interface with a 7 in. ControlPad featuring a capacitive touchscreen and durable, chemical-resistant glass surface.
  • Flexible configuration: one ControlPad can manage up to two Heating Modules, each accommodating up to two aluminum blocks (95 x 76 x 51 mm).
  • Independent control of temperature (up to 165°C) and timers for each Module.
  • Wide selection of interchangeable aluminum blocks suitable for various vials, tubes, and cuvettes.
  • Optional Transparent lid creates a warm air jacket, minimizing heat loss and ensuring uniform temperature distribution.
  • External Pt100 and Pt1000 probes allow precise temperature monitoring and control directly in the block or sample vessel.

Applications

  • DNA/RNA extraction and purification
  • Enzymatic reactions
  • Activation of cell cultures
  • DNA and protein denaturation
  • Assay preparation and clinical tests
  • Melting and boiling point determination
  • Culture media tests
  • Other analytical protocols

Accessory Needed

  • Unit must be ordered with block(s)

Specifications

Certifications/Compliance CE, UKCA
Controller Type ControlPad
Autoclavable Not Autoclavable
Temperature Range (Metric) 5°C to 165°C
Type Dry Block Heater
Display Digital
No. of Blocks 2 (sold separately)
Temperature Accuracy 0.5°C
Temperature Stability ±0.2°C at 37°C, ±0.4°C at 60°C, ±0.5°C at 165°C
Temperature Uniformity ±0.2°C at 37°C, ±0.4°C at 60°C, ±0.5°C at 165°C
Timer 1 min. to 99 hr. 59 min.
Warranty 3 Year
Dimensions (L x W x H) 228 x 350 x 92.5 mm
Weight (English) 4.6 lb.
Weight (Metric) 2.1 kg
Voltage 115/230 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.