missing translation for 'onlineSavingsMsg'
Learn More

Econazole, TRC

Product Code. 30388279
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388279 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388279 Supplier Toronto Research Chemicals Supplier No. TRCE3260031G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Econazole
CAS 27220-47-9
Molecular Formula C18 H15 Cl3 N2 O
Quantity 1 g
Synonym Econazole, 1-[(2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole, 1H-Imidazole, 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-, Imidazole, 1-[2,4-dichloro-β-[(p-chlorobenzyl)oxy]phenethyl]- (8CI), (±)-Econazole, 1-[2,4-Dichloro-β-[(p-chlorobenzyl)oxy]phenethyl]imidazole, Econazole, NSC 187789
InChI Formula InChI=1S/C18H15Cl3N2O/c19-14-3-1-13(2-4-14)11-24-18(10-23-8-7-22-12-23)16-6-5-15(20)9-17(16)21/h1-9,12,18H,10-11H2
SMILES Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)cc1
IUPAC Name 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole
Molecular Weight (g/mol) 381.68
Formula Weight 380.025
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.