Learn More
Socorex™ Ecostep Syringe Tips
Product Code. 15500754
Change view
Click to view available options
Volume (Metric):
50 to 500 μL
Unit Size:
Pack of 50
Product Code. Volume (Metric) Unit Size
15500754 50 to 500 μL Pack of 50
1 options
Product Code. 15500754 Supplier Socorex™ Supplier No. 316.050.9
Save Up to 60% on Liquid Handling
Use promo code 28028
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Volume (Metric) 50 to 500 μL
Filtered Non-filtered
Sterility Sterile
Accuracy <±0.8 to <±0.5%
Material Polypropylene