missing translation for 'onlineSavingsMsg'
Learn More

Edaravone, Tocris Bioscience™

Product Code. 30001426
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001426 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001426 Supplier Tocris Bioscience Supplier No. 0786/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anti-ischemic and antioxidant

A radical scavenger and antioxidant which is able to protect against the effects of ischemia, probably by inhibiting the lipoxygenase system. Protects against MPTP-induced neurotoxicity. Inhibits autophagy.
TRUSTED_SUSTAINABILITY

Specifications

CAS 89-25-8
Molecular Formula C10H10N2O
Quantity 50 mg
Synonym edaravone, 3-methyl-1-phenyl-2-pyrazolin-5-one, norphenazone, radicut, 1-phenyl-3-methyl-5-pyrazolone, developer z, methylphenylpyrazolone, norantipyrine, c.i. developer 1, phenylmethylpyrazolone
InChI Key QELUYTUMUWHWMC-UHFFFAOYSA-N
SMILES CC1=NN(C(=O)C1)C2=CC=CC=C2
IUPAC Name 5-methyl-2-phenyl-4H-pyrazol-3-one
Molecular Weight (g/mol) 174.203
PubChem CID 4021
ChEBI CHEBI:31530
Formula Weight 174.2
Percent Purity >99%
Chemical Name or Material Edaravone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.