Learn More
Ansell EDGE™ 48-706 Safety Gloves
Description
- Advanced durability: The nitrile foam-coated palm, EDGE™ 48-706 nitrile gloves offer a good grip and protection against cuts and abrasion in dry and slightly oily environments, making them ideal substitutes for heavy-duty leather and PU palm dipped, cut-resistant knitted gloves
- Increased protection: EDGE™ 48-706 safety gloves feature EN ISO 13997 level C and ANSI level A3 cut resistance, and meet EN 420 (general requirements) and EN 388 (mechanical hazards) safety standards
- Enhanced comfort: These breathable work gloves possess a seamless liner that elevates comfort without sacrificing dexterity, while delivering great value at an affordable cost
- Food handling compliance: Made of FDA-compliant materials, these protective gloves are safe for food handling and packaging
Specifications
Specifications
| Coating Material | Foam Nitrile |
| Color | Black |
| Cut Resistance Rating | Level C |
| Hand | Specific |
| Latex Free | Yes |
| Material | HPPE, Nylon, Polyester, Spandex, Foam Nitrile |
| Product Type | Industrial Gloves |
| Size | 6 |