missing translation for 'onlineSavingsMsg'
Learn More

Ansell EDGE™ 48-929 Double Layer Nitrile Coated Gloves

Product Code. 30259540 Shop All Ansell Products
Change view
Click to view available options
Size:
9
Unit Size:
12 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30259540 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30259540 Supplier Ansell Supplier No. 48929090
  • ONLINE EXCLUSIVE PRICE
    154.80 €
    98.42 € / 12 pairs
    Save 56.38 €
    36% Off
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Double layer NBR coating keeps hands dry and comfortable, in addition to enhancing grip capabilities when handling oily parts and tools.

  • Fully dipped NBR coating keeps hands dry and comfortable when handling oily parts and tools
  • Textured palm enhances grip capabilities
  • Seamless liner is comfortable and flexible
  • Elastic knitwrist ensures a secure fit
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Seamless Knit
Coating Material Nitrile
Color Black, Gray
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Fiberglass, Polyethylene, Spandex
Product Type Dip-coated Gloves
Size 9
EN388 Standard 4X43C
For Use With (Application) Aerospace, Automotive Assembly, Chemicals, Glass Manufacturing, Metal Fabrication, and Transportation
Glove Cuff Style Knit
Series 48-929
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.