Learn More
Ansell EDGE™ 48-969 Protective Gloves
Description
- Heightened grip: Featuring a fully NBR-coated seamless liner with a rough palm, EDGE™ 48-969 protective gloves offer a highly secure grip in oily environments, while protecting skin from injuries and oil contamination
- Optimized visibility: these bright yellow cut-resistant gloves ensure that hands remain highly visible, promoting safety even in low-light or dark conditions
- Enhanced defenses: EDGE™ 48-969 offers a high level of abrasion resistance and very high cut protection. In addition, the glove provides EN 407 level 1 contact heat protection, making it ideal for workers handling hot equipment, parts and objects.
Specifications
Specifications
| Coating Material | Nitrile |
| Color | Yellow |
| Cut Resistance Rating | Level F |
| Hand | Specific |
| Latex Free | Yes |
| Material | HPPE, Nitrile, Spandex, Nylon, Steel Fiber |
| Product Type | Industrial Gloves |
| Size | 10 |