Learn More
Contec™ Edgeless™ Sterile Mop Heads
BIOPHARMA
Product Code. 16420111 Shop All Contec Products
Change view
Click to view available options
Sterility:
Sterile
Description:
Edgeless Mop Head
Unit Size:
Pack of 20
Product Code. Sterility Description Unit Size
16420111 Sterile Edgeless Mop Head Pack of 20
1 options
Product Code. 16420111 Supplier Contec™ Supplier No. ELMOP IR
  • ONLINE EXCLUSIVE PRICE
    811.00 €
    687.64 € / Pack of 20
    Save 123.36 €
    15% Off
Estimated Shipment: 02-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Manufactured from a continuous tube of strong, durable 100% polyester knitted fabric. Edgeless™ Sterile Mop Heads are looped to form tubular mop strands that will not shred, break or deteriorate, eliminating contamination.
The Contec™ Edgeless™ Mopping System is the cleanest floor mopping system available for critical environments. The system is designed for semiconductor, pharmaceutical, biotech and medical device industries for lab clean up and process area cleaning. Mop heads are designed to meet general purpose cleaning needs in ISO Class 3 or less critical environments and are available in a wide variety of models to meet different cleaning needs. Edgeless™ or QuickConnect™ handles, frames, adaptors and bucket systems are available separately to complete the system.
  • Mop strands encapsulate liquids for maximum sorption; generate significantly lower particulate and extractable levels compared to conventional cotton string or nylon mops
  • Validated sterile per the AAMI/ISO Guidelines to a 10-6 Sterility Assurance Level (certificates of sterility available)
  • Mop heads laundered and packaged in an ISO Class 4 cleanroom
  • Resistant to chemicals, mold and mildew
  • Loops eliminate cut edges and provide improved cleanliness
  • Tail bands improved control and allow easy mopping around equipment legs and wheels
  • Available in three models:
    • Original
    • Textured--twisted strands increase sorbent capacity
    • Long textured--holds more liquid for mopping larger areas; lasts up to 10x longer than traditional mops; feature colored tail bands for area segregation to minimize cross-contamination
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mop Head
Color Code White
Sterility Sterile
Autoclavable Autoclavable
Disposable Limited-use
Description Edgeless Mop Head
Color-coded No
Material Polyester